Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)nicotinic acid 98+%

Supplier: AMBEED, INC
Warning

A1476081-100G A1476081-25G A1476081-500G A1476081-5G
76663-654EA 65 USD InStock
76663-654 76663-656 76663-658 76663-660
4-(Trifluoromethyl)nicotinic acid 98+%
4-(Trifluoromethyl)nicotinic acid
Precaución: for Research Use Only


Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
MDL Number: MFCD00082626
CAS Number: 158063-66-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty