Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2,6,10,10-Tetramethyl-1-oxaspiro[4.5]-5-decene 90% + (by GC, sum of isomers)

Supplier: AMBEED, INC
Warning

A1476147-100G A1476147-1G A1476147-25G A1476147-5G
76663-672EA 380 USD InStock
76663-672 76663-674 76663-676 76663-678
(±)-2,6,10,10-Tetramethyl-1-oxaspiro[4.5]-5-decene 90% + (by GC, sum of isomers)
(±)-2,6,10,10-Tetramethyl-1-oxaspiro[4.5]-5-decene
Precaución: for Research Use Only


Formula: C₁₃H₂₂O
MDL Number: MFCD00085214
CAS Number: 36431-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty