Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4-Isopropyl-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: AMBEED, INC
Warning

A1476165-100MG A1476165-1G A1476165-250MG
76663-722EA 42 USD InStock
76663-722 76663-724 76663-726
(R)-4-Isopropyl-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%
(R)-4-Isopropyl-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole
Precaución: for Research Use Only


Formula: C₁₂H₁₆N₂O
CAS Number: 2757083-00-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty