Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-((tert-Butoxycarbonyl)amino)-3-(2,4,5-trifluorophenyl)propanoic acid 98%

Supplier: AMBEED, INC
Warning

A1476692-1G A1476692-250MG A1476692-5G
76663-792EA 56 USD InStock
76663-792 76663-794 76663-796
(S)-2-((tert-Butoxycarbonyl)amino)-3-(2,4,5-trifluorophenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(2,4,5-trifluorophenyl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₆F₃NO₄
MW: 319.2764 g/mol
MDL Number: MFCD06659127
CAS Number: 324028-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty