Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzophenone 99%

Supplier: AMBEED, INC
Warning

A1477042-1000G A1477042-100G A1477042-25G A1477042-500G
76663-820EA 64 USD InStock
76663-820 76663-822 76663-824 76663-826
4-Hydroxybenzophenone 99%
4-Hydroxybenzophenone
Precaución: for Research Use Only


Formula: C₁₃H₁₀O₂
MDL Number: MFCD00002355
CAS Number: 1137-42-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty