Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-2-(4-propoxyphenyl)acetic acid 98+%

Supplier: AMBEED, INC
Warning

A1477062-100G A1477062-25G A1477062-500G
76663-860EA 59 USD InStock
76663-860 76663-862 76663-864
2-Hydroxy-2-(4-propoxyphenyl)acetic acid 98+%
2-Hydroxy-2-(4-propoxyphenyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
MDL Number: MFCD07368282
CAS Number: 79694-16-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty