Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorobenzophenone 98+%, (Chunks or pellets or flakes)

Supplier: AMBEED, INC
Warning

A1477288-100G A1477288-25G A1477288-500G
76663-990EA 21 USD InStock
76663-990 76663-992 76663-994
4-Chlorobenzophenone 98+%, (Chunks or pellets or flakes)
4-Chlorobenzophenone
Precaución: for Research Use Only


Formula: C₁₃H₉ClO
MDL Number: MFCD00000622
CAS Number: 134-85-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty