Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2-(2,6-diisopropylphenyl)isoindoline-1,3-dione 97%

Supplier: AMBEED, INC
Warning

A1481142-100MG A1481142-25MG A1481142-50MG
76664-038EA 242 USD InStock
76664-038 76664-040 76664-042
5-Amino-2-(2,6-diisopropylphenyl)isoindoline-1,3-dione 97%
5-Amino-2-(2,6-diisopropylphenyl)isoindoline-1,3-dione
Precaución: for Research Use Only


Formula: C₂₀H₂₂N₂O₂
MDL Number: MFCD03085319
CAS Number: 100823-03-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty