Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)benzene ≥97%, ee 99%

Supplier: AMBEED, INC
Warning

A1481266-100MG A1481266-1G A1481266-250MG
76664-056EA 48 USD InStock
76664-056 76664-058 76664-060
1,3-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)benzene ≥97%, ee 99%
1,3-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)benzene
Precaución: for Research Use Only


Formula: C₂₄H₂₀N₂O₂
MDL Number: MFCD31813234
CAS Number: 876953-19-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty