Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-nitrotoluene 98+%

Supplier: AMBEED, INC
Warning

A1481636-100G A1481636-1G A1481636-25G A1481636-5G
76664-120EA 111 USD InStock
76664-120 76664-122 76664-124 76664-126
3-Fluoro-5-nitrotoluene 98+%
3-Fluoro-5-nitrotoluene
Precaución: for Research Use Only


Formula: C7H6FNO2
MDL Number: MFCD06797943
CAS Number: 499-08-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty