Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-fluoro-6-nitrotoluene 98%

Supplier: AMBEED, INC
Warning

A1481643-100G A1481643-1G A1481643-25G A1481643-5G
76664-142EA 118 USD InStock
76664-142 76664-144 76664-146 76664-148
2-Bromo-4-fluoro-6-nitrotoluene 98%
2-Bromo-4-fluoro-6-nitrotoluene
Precaución: for Research Use Only


Formula: C₇H₅BrFNO₂
MDL Number: MFCD03094189
CAS Number: 502496-33-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty