Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-methylaniline 98%

Supplier: AMBEED, INC
Warning

A1481651-100G A1481651-25G A1481651-5G
76664-172EA 792 USD InStock
76664-172 76664-174 76664-176
2-Chloro-3-methylaniline 98%
2-Chloro-3-methylaniline
Precaución: for Research Use Only


Formula: C₇H₈ClN
MW: 141.598 g/mol
MDL Number: MFCD11113332
CAS Number: 29027-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty