Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,6-dichloroaniline 98%

Supplier: AMBEED, INC
Warning

A1481652-25G A1481652-5G A1481652-100G
76664-178EA 20 USD InStock
76664-178 76664-180 77624-348
4-Bromo-2,6-dichloroaniline 98%
4-Bromo-2,6-dichloroaniline
Precaución: for Research Use Only


Formula: C₆H₄BrCl₂N
MDL Number: MFCD00025162
CAS Number: 697-88-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty