Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Carbobenzoxyamino)valeric acid 98+%

Supplier: AMBEED, INC
Warning

A1481690-100G A1481690-25G A1481690-5G
76664-224EA 352 USD InStock
76664-224 76664-226 76664-228
5-(Carbobenzoxyamino)valeric acid 98+%
5-(Carbobenzoxyamino)valeric acid
Precaución: for Research Use Only


Formula: C₁₃H₁₇NO₄
MDL Number: MFCD00188458
CAS Number: 23135-50-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty