Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-3-amino-3-phenylpropionic acid 98+%

Supplier: AMBEED, INC
Danger

A1482461-1G A1482461-25G A1482461-5G
76664-286EA 28 USD InStock
76664-286 76664-288 76664-290
Fmoc-3-amino-3-phenylpropionic acid 98+%
Fmoc-3-amino-3-phenylpropionic acid
Precaución: for Research Use Only


Formula: C₂₄H₂₁NO₄
MDL Number: MFCD01863192
CAS Number: 180181-93-5
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty