Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(2-(((6-(trifluoromethyl)pyridin-2-yl)oxy)methyl)phenyl)acetate 97%

Supplier: AMBEED, INC
Warning

A1482498-100G A1482498-1G A1482498-25G A1482498-5G
76664-346EA 853 USD InStock
76664-346 76664-348 76664-350 76664-352
Methyl 2-(2-(((6-(trifluoromethyl)pyridin-2-yl)oxy)methyl)phenyl)acetate 97%
Methyl 2-(2-(((6-(trifluoromethyl)pyridin-2-yl)oxy)methyl)phenyl)acetate
Precaución: for Research Use Only


Formula: C₁₆H₁₄F₃NO₃
MW: 325.2825 g/mol
CAS Number: 187327-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty