Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-2,2'-thiodiglycolate 99%

Supplier: AMBEED, INC
Warning

A1483001-100G A1483001-25G A1483001-500G
76664-480EA 129 USD InStock
76664-480 76664-482 76664-484
Diethyl-2,2'-thiodiglycolate 99%
Diethyl-2,2'-thiodiglycolate
Precaución: for Research Use Only


Formula: C₈H₁₄O₄S
MW: 206.26 g/mol
MDL Number: MFCD00087932
CAS Number: 925-47-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty