Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminophenylboronic acid hydrochloride 98%

Supplier: AMBEED, INC
Warning

A1483723-100G A1483723-1G A1483723-25G A1483723-5G
76664-518EA 204 USD InStock
76664-518 76664-520 76664-522 76664-524
4-Aminophenylboronic acid hydrochloride 98%
4-Aminophenylboronic acid hydrochloride
Precaución: for Research Use Only


Formula: C₆H₉BClNO₂
MDL Number: MFCD03001333
CAS Number: 80460-73-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty