Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S)-2-Methylmorpholine hydrochloride 98+%

Supplier: AMBEED, INC
Warning

A1483771-1G A1483771-25G A1483771-5G
76664-622EA 49 USD InStock
76664-622 76664-624 76664-626
(2S)-2-Methylmorpholine hydrochloride 98+%
(2S)-2-Methylmorpholine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₁NO·ClH
MDL Number: MFCD11506701
CAS Number: 1147108-99-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty