Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indazole-4-carbaldehyde 99%

Supplier: AMBEED, INC
Warning

A1483772-100MG A1483772-1G A1483772-250MG A1483772-25G A1483772-5G
76664-628EA 7 USD InStock
76664-628 76664-630 76664-632 76664-634 76664-636
1H-Indazole-4-carbaldehyde 99%
1H-Indazole-4-carbaldehyde
Precaución: for Research Use Only


Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD06738279
CAS Number: 669050-70-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty