Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(1-BOC-pyrrolidin-3-yl)-acetic acid 98%

Supplier: AMBEED, INC
Danger

A1483782-1G A1483782-25G A1483782-5G A1483782-100G
76664-670EA 11 USD InStock
76664-670 76664-672 76664-674 77624-386
(R)-(1-BOC-pyrrolidin-3-yl)-acetic acid 98%
(R)-(1-BOC-pyrrolidin-3-yl)-acetic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₄
MW: 229.273 g/mol
MDL Number: MFCD05861547
CAS Number: 204688-60-8
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty