Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Methoxycyclohexan-1-amine 98+%

Supplier: AMBEED, INC
Warning

A1483789-100MG A1483789-1G A1483789-250MG A1483789-25G A1483789-5G
76664-706EA 6 USD InStock
76664-706 76664-708 76664-710 76664-712 76664-714
trans-4-Methoxycyclohexan-1-amine 98+%
trans-4-Methoxycyclohexan-1-amine
Precaución: for Research Use Only


Formula: C₇H₁₅NO
MDL Number: MFCD20278175
CAS Number: 121588-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty