Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Cyclopropylmethyl)pyrimidine-2,4(1H,3H)-dione 97%

Supplier: AMBEED, INC
Warning

A1483875-100MG A1483875-250MG
76664-762EA 23 USD InStock
76664-762 76664-764
1-(Cyclopropylmethyl)pyrimidine-2,4(1H,3H)-dione 97%
1-(Cyclopropylmethyl)pyrimidine-2,4(1H,3H)-dione
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O₂
MW: 166.17 g/mol
MDL Number: MFCD21105338
CAS Number: 1462994-89-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty