Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Oxo-2,3-dihydropyrimidin-4-yl)propionamide 97%

Supplier: AMBEED, INC
Warning

A1483883-100MG A1483883-1G A1483883-250MG
76664-780EA 27 USD InStock
76664-780 76664-782 76664-784
N-(2-Oxo-2,3-dihydropyrimidin-4-yl)propionamide 97%
N-(2-Oxo-2,3-dihydropyrimidin-4-yl)propionamide
Precaución: for Research Use Only


Formula: C₇H₉N₃O₂
CAS Number: 21261-08-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty