Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromo-1,4-difluorobenzene 98+%

Supplier: AMBEED, INC
Warning

A1485723-100G A1485723-1G A1485723-25G A1485723-5G
76664-868EA 236 USD InStock
76664-868 76664-870 76664-872 76664-874
2,3-Dibromo-1,4-difluorobenzene 98+%
2,3-Dibromo-1,4-difluorobenzene
Precaución: for Research Use Only


Formula: C₆H₂Br₂F₂
MW: 271.885 g/mol
MDL Number: MFCD12922620
CAS Number: 179737-33-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty