Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-chlorotoluene 98+%

Supplier: AMBEED, INC
Warning

A1485729-100G A1485729-25G A1485729-500G A1485729-5G
76664-892EA 45 USD InStock
76664-892 76664-894 76664-896 76664-898
2-Bromo-6-chlorotoluene 98+%
2-Bromo-6-chlorotoluene
Precaución: for Research Use Only


Formula: C₇H₆BrCl
MDL Number: MFCD00061121
CAS Number: 62356-27-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty