Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-chlorobenzotrifluoride 98+%

Supplier: AMBEED, INC
Warning

A1485744-100G A1485744-25G A1485744-500G
76664-960EA 28 USD InStock
76664-960 76664-962 76664-964 76664-966
2-Bromo-5-chlorobenzotrifluoride 98+%
2-Bromo-5-chlorobenzotrifluoride
Precaución: for Research Use Only


Formula: C₇H₃BrClF₃
MW: 259.45
MDL Number: MFCD00010308
CAS Number: 344-65-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty