Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-N-((R)-(5-(Diphenylphosphino)-9,9-dimethyl-9H-xanthen-4-yl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 95%

Supplier: AMBEED, INC
Warning

A1486510-100MG A1486510-250MG A1486510-50MG
76665-176EA 160 USD InStock
76665-176 76665-178 76665-180
(R)-N-((R)-(5-(Diphenylphosphino)-9,9-dimethyl-9H-xanthen-4-yl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 95%
(R)-N-((R)-(5-(Diphenylphosphino)-9,9-dimethyl-9H-xanthen-4-yl)(phenyl)methyl)-2-methylpropane-2-sulfinamide
Precaución: for Research Use Only


Formula: C₃₈H₃₈NO₂PS
MDL Number: MFCD33401725
CAS Number: 2757083-08-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty