Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabromobisphenol A bis(2-hydroxyethyl)ether 93+%

Supplier: AMBEED, INC
Warning

A1487502-100G A1487502-25G
76665-256EA 100 USD InStock
76665-256 76665-258 76665-260
Tetrabromobisphenol A bis(2-hydroxyethyl)ether 93+%
Tetrabromobisphenol A bis(2-hydroxyethyl)ether
Precaución: for Research Use Only


Formula: C₁₉H₂₀Br₄O₄
MDL Number: MFCD00059603
CAS Number: 4162-45-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty