Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Imidazole-4-carbodithioic acid 70+%

Supplier: AMBEED, INC
Warning

A1487581-1G A1487581-5G
76665-262EA 29 USD InStock
76665-262 76665-264
1H-Imidazole-4-carbodithioic acid 70+%
1H-Imidazole-4-carbodithioic acid
Precaución: for Research Use Only


Formula: C₄H₄N₂S₂
MDL Number: MFCD03428062
CAS Number: 84824-76-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty