Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[Ir(ppy)2([2,2'-bipyridine]-4-carboxylic acid)]Cl 98%

Supplier: AMBEED, INC
Warning

A1487625-100MG A1487625-1G A1487625-250MG A1487625-50MG
76665-272EA 151 USD InStock
76665-272 76665-274 76665-276 76665-278
[Ir(ppy)2([2,2'-bipyridine]-4-carboxylic acid)]Cl 98%
[Ir(ppy)2([2,2'-bipyridine]-4-carboxylic acid)]Cl
Precaución: for Research Use Only


Formula: C₃₃H₂₄ClIrN₄O₂
CAS Number: 2828447-19-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty