Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-((11bS)-2,6-Bis(4-methoxyphenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 98%

Supplier: AMBEED, INC
Warning

A1489600-100MG A1489600-1G A1489600-250MG A1489600-50MG
76665-486EA 177 USD InStock
76665-486 76665-488 76665-490 76665-492
N-((11bS)-2,6-Bis(4-methoxyphenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 98%
N-((11bS)-2,6-Bis(4-methoxyphenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide
Precaución: for Research Use Only


Formula: C₃₅H₂₅F₃NO₇PS
CAS Number: 2757287-59-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty