Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,22-Docosanedioic acid 98+%

Supplier: AMBEED, INC
Warning

A1490270-100G A1490270-1G A1490270-25G A1490270-5G
76665-664EA 977 USD InStock
76665-664 76665-666 76665-668 76665-670
1,22-Docosanedioic acid 98+%
1,22-Docosanedioic acid
Precaución: for Research Use Only


Formula: C₂₂H₄₂O₄
MDL Number: MFCD00002806
CAS Number: 505-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty