Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl Hydrogen Dodecanedioate ≥98%

Supplier: AMBEED, INC
Warning

A1490278-100G A1490278-1G
76665-680EA 1459 USD InStock
76665-680 76665-682
tert-Butyl Hydrogen Dodecanedioate ≥98%
tert-Butyl Hydrogen Dodecanedioate
Precaución: for Research Use Only


Formula: C₁₆H₃₀O₄
MDL Number: MFCD28098251
CAS Number: 234081-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty