Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl Hydrogen tetradecanedioate 98%

Supplier: AMBEED, INC
Warning

A1490280-1G A1490280-25G A1490280-5G
76665-684EA 16 USD InStock
76665-684 76665-686 76665-688
tert-Butyl Hydrogen tetradecanedioate 98%
tert-Butyl Hydrogen tetradecanedioate
Precaución: for Research Use Only


Formula: C₁₈H₃₄O₄
MDL Number: MFCD23136040
CAS Number: 234082-00-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty