Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

22-(tert-Butoxy)-22-oxodocosanoic acid 98+%

Supplier: AMBEED, INC
Warning

A1490292-100G A1490292-1G A1490292-250MG A1490292-25G A1490292-5G
76665-722EA 1808 USD InStock
76665-722 76665-724 76665-726 76665-728 76665-730
22-(tert-Butoxy)-22-oxodocosanoic acid 98+%
22-(tert-Butoxy)-22-oxodocosanoic acid
Precaución: for Research Use Only


Formula: C₂₆H₅₀O₄
MDL Number: MFCD30830215
CAS Number: 1642333-05-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty