Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

15-Bromopentadecanoic acid 98%

Supplier: AMBEED, INC
Warning

A1490306-25G A1490306-5G A1490306-1G
76665-740EA 895 USD InStock
76665-740 76665-742 77624-490
15-Bromopentadecanoic acid 98%
15-Bromopentadecanoic acid
Precaución: for Research Use Only


Formula: BrH₂C(CH₂)₁₃COOH
MW: 321.3
MDL Number: MFCD00210025
CAS Number: 56523-59-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty