Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Thietanamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A1490413-100MG A1490413-1G A1490413-250MG A1490413-5G
76665-752EA 33 USD InStock
76665-752 76665-754 76665-756 76665-758
3-Thietanamine hydrochloride 95%
3-Thietanamine hydrochloride
Precaución: for Research Use Only


Formula: C₃H₈ClNS
MW: 125.62 g/mol
MDL Number: MFCD19982300
CAS Number: 128861-78-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty