Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 97%

Supplier: AMBEED, INC
Warning

A1490462-1G A1490462-250MG A1490462-25G A1490462-5G A1490462-100MG
76665-766EA 75 USD InStock
76665-766 76665-768 76665-770 76665-772 77624-500
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 97%
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid
Precaución: for Research Use Only


Formula: C₇H₅BF₂O₄
MW: 201.92 g/mol
MDL Number: MFCD08706236
CAS Number: 190903-71-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty