Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopent-2-enone 98+%

Supplier: AMBEED, INC
Warning

A1490474-100MG A1490474-1G A1490474-250MG
76665-774EA 45 USD InStock
76665-774 76665-776 76665-778
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopent-2-enone 98+%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopent-2-enone
Precaución: for Research Use Only


Formula: C₁₁H₁₇BO₃
MW: 208.06 g/mol
MDL Number: MFCD22683474
CAS Number: 1370008-65-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty