Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Ir[Me(Me)ppy]2(4,4'-dCF3bpy))PF6 97%

Supplier: AMBEED, INC
Warning

A1490576-100MG A1490576-250MG A1490576-25MG A1490576-50MG
76665-788EA 88 USD InStock
76665-788 76665-790 76665-792 76665-794
(Ir[Me(Me)ppy]2(4,4'-dCF3bpy))PF6 97%
(Ir[Me(Me)ppy]2(4,4'-dCF3bpy))PF6
Precaución: for Research Use Only


Formula: C₃₈H₃₀F₁₂IrN₄P
CAS Number: 2829282-26-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty