Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4S,4'S)-2,2'-((Phenylphosphinediyl)bis(2,1-phenylene))bis(4-isobutyl-4,5-dihydrooxazole) 97%

Supplier: AMBEED, INC
Warning

A1493550-100MG A1493550-250MG A1493550-50MG
76665-960EA 143 USD InStock
76665-960 76665-962 76665-964
(4S,4'S)-2,2'-((Phenylphosphinediyl)bis(2,1-phenylene))bis(4-isobutyl-4,5-dihydrooxazole) 97%
(4S,4'S)-2,2'-((Phenylphosphinediyl)bis(2,1-phenylene))bis(4-isobutyl-4,5-dihydrooxazole)
Precaución: for Research Use Only


Formula: C₃₂H₃₇N₂O₂P
CAS Number: 2757083-14-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty