Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2R)-1,2-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)cyclohexane 97%

Supplier: AMBEED, INC
Warning

A1493984-100MG A1493984-1G A1493984-250MG
76666-136EA 123 USD InStock
76666-136 76666-138 76666-140
(1R,2R)-1,2-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)cyclohexane 97%
(1R,2R)-1,2-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)cyclohexane
Precaución: for Research Use Only


Formula: C₂₄H₂₆N₂O₂
CAS Number: 2757083-39-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty