Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenylphosphine oxide ≥98% (Loss on drying ≤1%)

Supplier: AMBEED, INC
Warning

A1494181-100G A1494181-25G A1494181-5G
76666-230EA 29 USD InStock
76666-230 76666-232 76666-234
Diphenylphosphine oxide ≥98% (Loss on drying ≤1%)
Diphenylphosphine oxide
Precaución: for Research Use Only


Formula: C₁₂H₁₁OP
MDL Number: MFCD00002079
CAS Number: 4559-70-0
Order Now
Specification Test Results
Loss on drying ≤ 1%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty