Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Amino-2-methylphenyl)dimethylphosphine oxide 95%

Supplier: AMBEED, INC
Warning

A1495224-100MG A1495224-250MG A1495224-50MG
76666-300EA 179 USD InStock
76666-300 76666-302 76666-304
(4-Amino-2-methylphenyl)dimethylphosphine oxide 95%
(4-Amino-2-methylphenyl)dimethylphosphine oxide
Precaución: for Research Use Only


Formula: C₉H₁₄NOP
CAS Number: 2361948-73-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty