Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Amino-3-methoxyphenyl)dimethylphosphine oxide 95%

Supplier: AMBEED, INC
Warning

A1495225-100MG A1495225-1G A1495225-250MG A1495225-50MG
76666-306EA 163 USD InStock
76666-306 76666-308 76666-310 76666-312
(4-Amino-3-methoxyphenyl)dimethylphosphine oxide 95%
(4-Amino-3-methoxyphenyl)dimethylphosphine oxide
Precaución: For research use only.


Formula: C₉H₁₄NO₂P
MDL Number: MFCD30303983
CAS Number: 1197956-03-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty