Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 6-chloropyridine-3,4-dicarboxylate 95+%

Supplier: AMBEED, INC
Danger

A1495410-100MG A1495410-1G A1495410-250MG
76666-348EA 134 USD InStock
76666-348 76666-350 76666-352
Dimethyl 6-chloropyridine-3,4-dicarboxylate 95+%
Dimethyl 6-chloropyridine-3,4-dicarboxylate
Precaución: for Research Use Only


Formula: C₉H₈ClNO₄
MW: 229.6171 g/mol
MDL Number: MFCD28972751
CAS Number: 1189570-18-0
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty