Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[4,4'-Bis-trifluoromethyl]bis[(4-fluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%

Supplier: AMBEED, INC
Warning

A1495450-100MG A1495450-1G A1495450-250MG
76666-370EA 105 USD InStock
76666-370 76666-372 76666-374
[4,4'-Bis-trifluoromethyl]bis[(4-fluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%
[4,4'-Bis-trifluoromethyl]bis[(4-fluorophenyl)pyridine]iridium(III) hexafluorophosphate
Precaución: for Research Use Only


Formula: C₃₄H₂₀F₁₄IrN₄P
MW: 973.7178 g/mol
CAS Number: 2757085-01-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty