Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[4,4'-Bis-trifluoromethyl]bis[(2,4-difluoro-2-(5-methyl-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate 97%

Supplier: AMBEED, INC
Warning

A1495454-1G A1495454-250MG A1495454-50MG
76666-400EA 1152 USD InStock
76666-400 76666-402 76666-404
[4,4'-Bis-trifluoromethyl]bis[(2,4-difluoro-2-(5-methyl-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate 97%
[4,4'-Bis-trifluoromethyl]bis[(2,4-difluoro-2-(5-methyl-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate
Precaución: for Research Use Only


Formula: C₃₆H₂₂F₁₆IrN₄P
CAS Number: 2757085-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty