Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dioxoisoindolin-2-yl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate 97%

Supplier: AMBEED, INC
Warning

A1496217-1G A1496217-250MG A1496217-5G
76666-596EA 83 USD InStock
76666-596 76666-598 76666-600
1,3-Dioxoisoindolin-2-yl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate 97%
1,3-Dioxoisoindolin-2-yl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate
Precaución: for Research Use Only


Formula: C₂₃H₂₅NO₅
CAS Number: 1788861-30-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty